Learn More
KRT23, Mouse, Clone: AT2F6, Abnova™
Mouse monoclonal antibody raised against partial recombinant KRT23.
Brand: Abnova MAB11191.100uL
Description
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21. [provided by RefSeq
Specifications
| KRT23 | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against partial recombinant KRT23. | |
| In Phosphate-Buffered Saline (pH 7.4) with 0.09% sodium azide. | |
| CK23/DKFZp434G032/HAIK1/K23/MGC26158 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 25984 | |
| Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles. | |
| IgG1 κ |
| ELISA, Western Blot | |
| AT2F6 | |
| Unconjugated | |
| Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
| KRT23 | |
| KRT23 | |
| Recombinant protein corresponding to amino acids 271-422 of human KRT23. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.