missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT8/KRT18, Mouse, Clone: CK8+18 207, Abnova™
Mouse monoclonal antibody raised against KRT8/KRT18.
Brand: Abnova MAB6191.500uL
This item is not returnable.
View return policy
Specifications
| KRT8/KRT18 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| CARD2/CK8/CYK8/K2C8/K8/KO | |
| Mouse | |
| 500 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| CK8+18 207 | |
| Mouse monoclonal antibody raised against KRT8/KRT18. | |
| In PBS, pH 7.4 (1% BSA, 0.05% sodium azide) | |
| KRT8 | |
| Human KRT8/KRT18. | |
| RUO | |
| 3856/3875 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction