missing translation for 'onlineSavingsMsg'
Learn More
Learn More
La Crosse virus, Mouse, Clone: 9E7.2, Abnova™
Mouse monoclonal antibody raised against La Crosse virus.
Brand: Abnova MAB8234.100ug
This item is not returnable.
View return policy
Specifications
| La Crosse virus | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 9E7.2 | |
| Mouse monoclonal antibody raised against La Crosse virus. | |
| In PBS | |
| Native La Crosse Virus (LACV) from E6 Vero cells. | |
| RUO | |
| G2 envelope glycoprotein (35kD); virus-neutralizing | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction