missing translation for 'onlineSavingsMsg'
Learn More

LAMP2, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant LAMP2.

Brand:  Abnova H00003920-A01.50uL

Product Code. 16180575

  • € 242.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is a member of a family of membrane glycoproteins. This glycoprotein provides selectins with carbohydrate ligands. It may play a role in tumor cell metastasis. It may also function in the protection, maintenance, and adhesion of the lysosome. Alternative splicing of this gene results in multiple transcript variants encoding distinct proteins. [provided by RefSeq

Sequence: ELNLTDSENATCLYAKWQMNFTVRYETTNKTYKTVTISDHGTVTYNGSICGDDQNGPKIAVQFGPGFSWIANFTKAASTYSIDSVSFSYNTGDNTTFP
Specifications

Specifications

LAMP2
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant LAMP2.
LAMP2
CD107b/LAMPB/LGP110
Mouse
50 μL
Primary
Human
Antisera
ELISA, Western Blot
Unconjugated
50% glycerol
NM_013995
LAMP2
LAMP2 (NP_054701, 30 a.a. ∽ 127 a.a) partial recombinant protein with GST tag.
RUO
3920
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only