Learn More
LDHA, Mouse, Clone: 20, Abnova™
Mouse monoclonal antibody raised against full-length recombinant LDHA.
Brand: Abnova MAB9910.100ug
Description
The protein encoded by this gene catalyzes the conversion of L-lactate and NAD to pyruvate and NADH in the final step of anaerobic glycolysis. The protein is found predominantly in muscle tissue and belongs to the lactate dehydrogenase family. Mutations in this gene have been linked to exertional myoglobinuria. Multiple transcript variants encoding different isoforms have been found for this gene. The human genome contains several nontranscribed pseudogenes of this gene. [provided by RefSeq
Specifications
| LDHA | |
| Monoclonal | |
| Unconjugated | |
| Western blot (1:500) The optimal working dilution should be determined by the end user. | |
| LDHA | |
| LDH-M/LDH1/PIG19 | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 3939 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 20 | |
| Mouse monoclonal antibody raised against full length recombinant LDHA. | |
| In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
| NM_005566.3 | |
| LDHA | |
| Recombinant protein corresponding to full length human LDHA. | |
| 100 μg | |
| Primary | |
| It can expression in HepG2 whole cell lysate. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.