missing translation for 'onlineSavingsMsg'
Learn More

Lime1, Mouse, Clone: mLIME-05, Abnova™

Product Code. 16067416
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16067416 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16067416 Supplier Abnova Supplier No. MAB8509.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against partial recombiant Lime1.

Specifications

Antigen Lime1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone mLIME-05
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombiant Lime1.
Dilution Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Lime1
Gene Alias 2810038K19Rik/LIME
Gene Symbols Lime1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 86-202 of mouse Lime1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72699
Test Specificity This antibody is specific to cytoplasmic domain of Lime1.
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.