Learn More
LITAF, Mouse, Clone: AT1F9, Abnova™
Mouse monoclonal antibody raised against full-length recombinant LITAF.
Brand: Abnova MAB5642.100uL
Description
Lipopolysaccharide is a potent stimulator of monocytes and macrophages, causing secretion of tumor necrosis factor-alpha (TNF-alpha) and other inflammatory mediators. This gene encodes lipopolysaccharide-induced TNF-alpha factor, which is a DNA-binding protein and can mediate the TNF-alpha expression by direct binding to the promoter region of the TNF-alpha gene. The transcription of this gene is induced by tumor suppresor p53 and has been implicated in the p53-induced apoptotic pathway. Mutations in this gene cause Charcot-Marie-Tooth disease type 1C (CMT1C) and may be involved in the carcinogenesis of extramammary Paget's disease (EMPD). Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]
Specifications
| LITAF | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| LITAF | |
| LITAF | |
| Recombinant protein corresponding to full length human LITAF. | |
| RUO | |
| 9516 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| ELISA, Western Blot | |
| AT1F9 | |
| Mouse monoclonal antibody raised against full length recombinant LITAF. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| FLJ38636/MGC116698/MGC116700/MGC116701/MGC125274/MGC125275/MGC125276/PIG7/SIMPLE/TP53I7 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.