Learn More
LPL, Mouse, Clone: 2C5, Abnova™
Mouse monoclonal antibody raised against partial recombinant LPL.
Brand: Abnova MAB10791.100uL
Description
LPL encodes lipoprotein lipase, which is expressed in heart, muscle, and adipose tissue. LPL functions as a homodimer, and has the dual functions of triglyceride hydrolase and ligand/bridging factor for receptor-mediated lipoprotein uptake. Severe mutations that cause LPL deficiency result in type I hyperlipoproteinemia, while less extreme mutations in LPL are linked to many disorders of lipoprotein metabolism. [provided by RefSeq
Specifications
| LPL | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| LPL | |
| LPL | |
| Recombinant protein corresponding to human LPL. | |
| RUO | |
| 4023 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Western Blot | |
| 2C5 | |
| Mouse monoclonal antibody raised against partial recombinant LPL. | |
| In ascites (0.03% sodium azide) | |
| HDLCQ11/LIPD | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.