Learn More
LTF, Mouse, TRITC, Clone: NI 25, Abnova™
Mouse monoclonal antibody raised against LTF.
Brand: Abnova MAB10072.200ug
Description
This gene is a member of the transferrin family of genes and its protein product is found in the secondary granules of neutrophils. The protein is a major iron-binding protein in milk and body secretions with an antimicrobial activity, making it an important component of the nonspecific immune system. The protein demonstrates a broad spectrum of properties, including regulation of iron homeostasis, host defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation and protection against cancer development and metastasis. [provided by RefSeq
Specifications
| LTF | |
| Monoclonal | |
| TRITC | |
| Immunohistochemistry (1:10-1:100) The optimal working dilution should be determined by the end user. | |
| LTF | |
| LTF | |
| Purified LTF from human milk. | |
| RUO | |
| 4057 | |
| Human | |
| Lyophilized |
| Immunofluorescence, Immunohistochemistry | |
| NI 25 | |
| Mouse monoclonal antibody raised against LTF. | |
| Lyophilized from PBS, pH 7.2 | |
| GIG12/HLF2/LF | |
| Mouse | |
| 200 μg | |
| Primary | |
| The antiserum does not react with any other component of the human milk or plasma protein as tested. This antiserum has not been tested for cross-reactivity with other species. | |
| Store at 4°C on dry atmosphere. After reconstitution with 0.5mL deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.