Learn More
MAFA, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MAFA.
Brand: Abnova H00389692-A01.50uL
Description
MAFA is a transcription factor that binds RIPE3b, a conserved enhancer element that regulates pancreatic beta cell-specific expression of the insulin gene (INS; MIM 176730) (Olbrot et al., 2002 [PubMed 12011435]).[supplied by OMIM
Sequence: LEERFSDDQLVSMSVRELNRQLRGFSKEEVIRLKQKRRTLKNRGYAQSCRFKRVQQRHILESEKCQLQSQVEQLKLEVGRLAKERDLSpecifications
| MAFA | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MAFA. | |
| MAFA | |
| RIPE3b1/hMafA | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_201589 | |
| MAFA | |
| MAFA (NP_963883, 222 a.a. ∽ 308 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 389692 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only