Learn More
MAP4K4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MAP4K4.
Brand: Abnova H00009448-A01.50uL
Description
The protein encoded by this gene is a member of the serine/threonine protein kinase family. This kinase has been shown to specifically activate MAPK8/JNK. The activation of MAPK8 by this kinase is found to be inhibited by the dominant-negative mutants of MAP3K7/TAK1, MAP2K4/MKK4, and MAP2K7/MKK7, which suggests that this kinase may function through the MAP3K7-MAP2K4-MAP2K7 kinase cascade, and mediate the TNF-alpha signaling pathway. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: VHPALQRPAEPQVQWSHLASLKNNVSPVSRSHSFSDPSPKFAHHHLRSQDPCPPSRSEVLSQSSDSKSEAPDPTQKAWSRSDSDEVPPRVPVRTTSRSPVSpecifications
| MAP4K4 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MAP4K4. | |
| MAP4K4 | |
| FLH21957/FLJ10410/FLJ20373/FLJ90111/HGK/KIAA0687/NIK | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_145686 | |
| MAP4K4 | |
| MAP4K4 (NP_663719, 611 a.a. ∽ 710 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 9448 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only