Learn More
MAPKAPK5, Mouse, Clone: 7H10B4, Abnova™
Mouse monoclonal antibody raised against recombinant MAPKAPK5.
Brand: Abnova MAB10404.100uL
Description
The protein encoded by this gene is a member of the serine/threonine kinase family. In response to cellular stress and proinflammatory cytokines, this kinase is activated through its phosphorylation by MAP kinases including MAPK1/ERK, MAPK14/p38-alpha, and MAPK11/p38-beta. In vitro, this kinase phosphorylates heat shock protein HSP27 at its physiologically relevant sites. Two alternately spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq
Specifications
| MAPKAPK5 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| MAPKAPK5 | |
| MAPKAPK5 | |
| Recombinant protein corresponding to human MAPKAPK5. | |
| RUO | |
| 8550 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 7H10B4 | |
| Mouse monoclonal antibody raised against recombinant MAPKAPK5. | |
| In ascites (0.03% sodium azide) | |
| PRAK | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.