missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mdc1, Mouse, Biotin, Clone: P2B11, Abnova™
Mouse monoclonal antibody raised against recombinant partial Mdc1.
Brand: Abnova MAB11471.100ug
This item is not returnable.
View return policy
Specifications
| Mdc1 | |
| Monoclonal | |
| Biotin | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Mdc1 | |
| Mdc1 | |
| Recombinant GST fusion protein corresponding to C-terminus of mouse Mdc1. | |
| 100 μg | |
| Primary | |
| Bovine, Chimpanzee, Human, Mouse | |
| Liquid |
| Immunofluorescence, Western Blot | |
| P2B11 | |
| Mouse monoclonal antibody raised against recombinant partial Mdc1. | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| 6820401C03/AA413496/Nfbd1/mKIAA0170 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 240087 | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction