Learn More
MKLN1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MKLN1.
Brand: Abnova H00004289-A01.50uL
Description
Muskelin is an intracellular protein that acts as a mediator of cell spreading and cytoskeletal responses to the extracellular matrix component thrombospondin I (MIM 188060) (Adams et al., 1998 [PubMed 9724633]).[supplied by OMIM
Sequence: RHCKYLIRKHRFEEKAQVDPLSALKYLQNDLYITVDHSDPEETKEFQLLASALFKSGSDFTALGFSDVDHTYAQRTQLFDTLVNFFPDSMTPPKGNLVDLITLSpecifications
| MKLN1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MKLN1. | |
| MKLN1 | |
| FLJ11162/TWA2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_013255 | |
| MKLN1 | |
| MKLN1 (NP_037387, 633 a.a. ∽ 735 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 4289 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only