Learn More
MLC1SA, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MLC1SA.
Brand: Abnova H00140465-A01.50uL
Description
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in both slow-twitch skeletal muscle and in nonmuscle tissue. [provided by RefSeq
Sequence: LGNPKSDELKSRRVDFETFLPMLQAVAKNRGQGTYEDYLEGFRVFDKEGNGKVMGAELRHVLTTLGEKMTEEEVETVLAGHEDSNGCINYEAFLKHILSVSpecifications
| MLC1SA | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MLC1SA. | |
| MYL6B | |
| MLC1SA | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002475 | |
| MYL6B | |
| MLC1SA (NP_002466, 109 a.a. ∽ 208 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 140465 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only