missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mme, Mouse, Clone: LT10, Abnova™
Mouse monoclonal antibody raised against native Mme.
Brand: Abnova MAB3867.100ug
This item is not returnable.
View return policy
Specifications
| Mme | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mme | |
| Mme | |
| Native purified Mme from mouse NALM-6 leukemia pre-B cell line. | |
| RUO | |
| 17380 | |
| Human, Mouse | |
| Liquid |
| Flow Cytometry | |
| LT10 | |
| Mouse monoclonal antibody raised against native Mme. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| 6030454K05Rik/C85356/CALLA/CD10/NEP | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts with CD10 antigen (CALLA-Common acute lymphatic leukemiaantigen), a 100 KDa type II integral membrane protein. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction