Learn More
MMP9, Mouse, Clone: 2C3, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of MMP9.
Brand: Abnova MAB1952.200ug
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The enzyme encoded by this gene degrades type IV and V collagens. Studies in rhesus monkeys suggest that the enzyme is involved in IL-8-induced mobilization of hematopoietic progenitor cells from bone marrow, and murine studies suggest a role in tumor-associated tissue remodeling. [provided by RefSeq
Specifications
| MMP9 | |
| Monoclonal | |
| Unconjugated | |
| Western blot (1 to 2 ug/mL) Immunohistochemistry on formalin to fixed/paraffin to embedded tissues (1 to 5 ug/mL) The optimal working dilution should be determined by the end user. | |
| MMP9 | |
| MMP9 | |
| A synthetic peptide corresponding to C-terminus of human MMP9. | |
| RUO | |
| 4318 | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 2C3 | |
| Mouse monoclonal antibody raised against synthetic peptide of MMP9. | |
| In PBS (0.08% sodium azide) | |
| CLG4B/GELB/MMP-9 | |
| Mouse | |
| 200 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.