Learn More
MS4A1, Mouse, FITC, Clone: LT20, Abnova™
Mouse monoclonal antibody raised against native MS4A1.
Brand: Abnova MAB4385.100Reactions
Description
This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. This gene encodes a B-lymphocyte surface molecule which plays a role in the development and differentiation of B-cells into plasma cells. This family member is localized to 11q12, among a cluster of family members. Alternative splicing of this gene results in two transcript variants which encode the same protein. [provided by RefSeq
Specifications
| MS4A1 | |
| Monoclonal | |
| FITC | |
| Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user. | |
| MS4A1 | |
| MS4A1 | |
| Native purified MS4A1 from normal human lymphocytes from lymph node. | |
| RUO | |
| 931 | |
| Human | |
| Liquid |
| Flow Cytometry | |
| LT20 | |
| Mouse monoclonal antibody raised against native MS4A1. | |
| In PBS (0.2% BSA, 0.09% sodium azide) | |
| B1/Bp35/CD20/LEU-16/MGC3969/MS4A2/S7 | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| This antibody reacts with CD20 (Bp35), a 33-37 KDa non-glycosylated membrane receptor with four transmembrane domains, expressed on B lymphocytes (it is lost on plasma cells), follicular dendritic cells, and at low levels on peripheral blood T lymphocytes. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.