Learn More
MSR1, Mouse, Clone: SRA-E5, Abnova™
Mouse monoclonal antibody raised against recombinant MSR1.
Brand: Abnova MAB1710.50ug
Description
This gene encodes the class A macrophage scavenger receptors, which include three different types (1, 2, 3) generated by alternative splicing of this gene. These receptors or isoforms are macrophage-specific trimeric integral membrane glycoproteins and have been implicated in many macrophage-associated physiological and pathological processes including atherosclerosis, Alzheimer's disease, and host defense. The isoforms type 1 and type 2 are functional receptors and are able to mediate the endocytosis of modified low density lipoproteins (LDLs). The isoform type 3 does not internalize modified LDL (acetyl-LDL) despite having the domain shown to mediate this function in the types 1 and 2 isoforms. It has an altered intracellular processing and is trapped within the endoplasmic reticulum, making it unable to perform endocytosis. The isoform type 3 can inhibit the function of isoforms type 1 and type 2 when co-expressed, indicating a dominant negative effect and suggesting a mechanism for regulation of scavenger receptor activity in macrophages. [provided by RefSeq]
Specifications
| MSR1 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (10 ug/mL) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| MSR1 | |
| MSR1 | |
| Recombinant protein corresponding to human MSR1. | |
| RUO | |
| 4481 | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| SRA-E5 | |
| Mouse monoclonal antibody raised against recombinant MSR1. | |
| In PBS (0.1% proclin, 2.0% Block Ace) | |
| CD204/SCARA1/SR-A/phSR1/phSR2 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.