missing translation for 'onlineSavingsMsg'
Learn More

MUC16, Mouse, Biotin, Clone: X306, Abnova™

Product Code. 16096856
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16096856 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16096856 Supplier Abnova Supplier No. MAB5523.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against native MUC16.

Specifications

Antigen MUC16
Applications Western Blot
Classification Monoclonal
Clone X306
Conjugate Biotin
Description Mouse monoclonal antibody raised against native MUC16.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (0.05% sodium azide)
Gene MUC16
Gene Alias CA125/FLJ14303
Gene Symbols MUC16
Host Species Mouse
Immunogen Native purified human MUC16.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94025
Test Specificity Specific to group A.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.