Learn More
MUC2, Mouse, Clone: SPM296, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of MUC2.
Brand: Abnova MAB12113.100ug
Description
This gene encodes a member of the mucin protein family. Mucins are high molecular weight glycoproteins produced by many epithelial tissues. The protein encoded by this gene is secreted and forms an insoluble mucous barrier that protects the gut lumen. The protein polymerizes into a gel of which 80% is composed of oligosaccharide side chains by weight. The protein features a central domain containing tandem repeats rich in threonine and proline that varies between 50 and 115 copies in different individuals. Alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known. [provided by RefSeq
Sequence: KYPTTTPISTTTMVTPTPTPTGTQTPTTTSpecifications
| MUC2 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (0.5-1 ug/mL) Western blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| MUC2 | |
| MUC2 | |
| A synthetic peptide of 29 amino acids corresponding to human MUC2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| SPM296 | |
| Mouse monoclonal antibody raised against synthetic peptide of MUC2. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| MLP/SMUC | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 4583 | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.