missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloid Specific Antigen, Mouse, Clone: BM-2, Abnova™
Mouse monoclonal antibody raised against native Myeloid Specific Antigen.
Brand: Abnova MAB5183.100ug
This item is not returnable.
View return policy
Specifications
| Myeloid Specific Antigen | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Frozen) | |
| BM-2 | |
| Mouse monoclonal antibody raised against native Myeloid Specific Antigen. | |
| In PBS (0.05% sodium azide) | |
| Native purified from pokeweed mitogen stimulated human peripheral blood lymphocyte nuclei. | |
| RUO | |
| BM-2 recognizes a cytoplasmic antigen expressed in mature human granulocytes (polys) residing in lymphoid and non-lymphoid tissues. It does not react with any other cell type in human tissues. | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction