missing translation for 'onlineSavingsMsg'
Learn More

NANOG, Mouse, Clone: 5A10, Abnova™

Product Code. 16170500
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16170500 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16170500 Supplier Abnova Supplier No. MAB2042.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against partial recombinant NANOG.

Specifications

Antigen NANOG
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5A10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant NANOG.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene NANOG
Gene Symbols NANOG
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-154 of human NANOG.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79923
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.