Learn More
NBR1, Mouse, Clone: 3D3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NBR1.
Brand: Abnova H00004077-M03A.200uL
Description
The protein encoded by this gene was originally identified as an ovarian tumor antigen monitored in ovarian cancer. The encoded protein contains a B-box/coiled coil motif, which is present in many genes with transformation potential, but the function of this protein is unknown. This gene is located on a region of chromosome 17q21.1 that is in close proximity to tumor suppressor gene BRCA1. Three alternatively spliced variants encoding the same protein have been identified for this gene. [provided by RefSeq
Sequence: EPQVTLNVTFKNEIQSFLVSDPENTTWADIEAMVKVSFDLNTIQIKYLDEENEEVSINSQGEYEEALKMAVKQGNQLQMQVHEGHHVVDEAPPPVSpecifications
| NBR1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_005899 | |
| NBR1 | |
| NBR1 (NP_005890,2 a.a. ∽ 96 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4077 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 3D3 | |
| Mouse monoclonal antibody raised against a partial recombinant NBR1. | |
| NBR1 | |
| 1A1-3B/KIAA0049/M17S2/MIG19 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.