missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAM1, Mouse, Clone: ANC7C7, Abnova™
Mouse monoclonal antibody raised against recombinant NCAM1.
Brand: Abnova MAB6950.100ug
This item is not returnable.
View return policy
Specifications
| NCAM1 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| NCAM1 | |
| NCAM1 | |
| Recombinant protein corresponding to human NCAM1. | |
| RUO | |
| 4684 | |
| Store at 4°C. | |
| IgG1 κ |
| ELISA, Flow Cytometry | |
| ANC7C7 | |
| Mouse monoclonal antibody raised against recombinant NCAM1. | |
| In 50mM sodium phosphate buffer, 100mM potassium chloride, 150mM NaCl, pH 7.5 (0.5mg/mL gentamicin sulfate) | |
| CD56/MSK39/NCAM | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction