missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ncan, Mouse, Clone: 650.24, Abnova™
Mouse monoclonal antibody raised against native Ncan.
Brand: Abnova MAB2432.100uL
This item is not returnable.
View return policy
Specifications
| Ncan | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Ncan | |
| Ncan | |
| Native purified rat Ncan. | |
| RUO | |
| 58982 | |
| Rat | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 650.24 | |
| Mouse monoclonal antibody raised against native Ncan. | |
| In PBS (0.09% sodium azide) | |
| Cspg3 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Neurocan core protein, also reacts with Ncan fragment. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction