missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEDD8, Mouse, Clone: 1A7, Abnova™
Mouse monoclonal antibody raised against partial recombinant NEDD8.
Brand: Abnova MAB10825.100ug
This item is not returnable.
View return policy
Specifications
| NEDD8 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| NEDD8 | |
| NEDD8 | |
| Recombinant protein corresponding to human NEDD8. | |
| RUO | |
| 4738 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1A7 | |
| Mouse monoclonal antibody raised against partial recombinant NEDD8. | |
| In PBS (0.5% protein stabilizer, 0.05% sodium azide) | |
| FLJ43224/MGC104393/MGC125896/MGC125897/Nedd-8 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction