missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEUROD6, Mouse, Clone: 4G11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NEUROD6.
Brand: Abnova H00063974-M02A.200uL
This item is not returnable.
View return policy
Description
NEUROD6 is a member of the NEUROD (NEUROD1; MIM 601724) family of basic helix-loop-helix (bHLH) transcription factors (Guo et al., 2002 [PubMed 12357074]).[supplied by OMIM
Sequence: GQGGEAAHHTRSPYSTFYPPYHSPELTTPPGHGTLDNSKSMKPYNYCSAYESFYESTSPECASPQFEGPLSPPPINYNGIFSLKQEETLDYGKNYNYGMHSpecifications
| NEUROD6 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_022728 | |
| NEUROD6 | |
| NEUROD6 (NP_073565.2,191 a.a. ∽ 290 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 63974 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 4G11 | |
| Mouse monoclonal antibody raised against a partial recombinant NEUROD6. | |
| NEUROD6 | |
| Atoh2/MATH2/Math-2/NEX1M/bHLHa2 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction