Learn More
NLGN3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant NLGN3.
Brand: Abnova H00054413-A01.50uL
Description
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene, but their full length sequences have not been determined. [provided by RefSeq
Sequence: PRVRDHYRATKVAFWKHLVPHLYNLHDMFHYTSTTTKVPPPDTTHSSHITRRPNGKTWSTKRPAISPAYSNENAQGSWNGDQDAGPLLVENPRDYSTELSSpecifications
| NLGN3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant NLGN3. | |
| NLGN3 | |
| ASPGX1/AUTSX1/HNL3/KIAA1480 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_018977 | |
| NLGN3 | |
| NLGN3 (NP_061850, 590 a.a. ∽ 689 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 54413 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only