Learn More
PAIP1, Mouse, Clone: 7E7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PAIP1.
Brand: Abnova H00010605-M02A.200uL
Description
The protein encoded by this gene interacts with poly(A)-binding protein and with the cap-binding complex eIF4A. It is involved in translational initiation and protein biosynthesis. Overexpression of this gene in COS7 cells stimulates translation. Alternative splicing occurs at this locus and three transcript variants encoding three distinct isoforms have been identified. [provided by RefSeq
Sequence: YPTLSEYVQDFLNHLTEQPGSFETEIEQFAETLNGCVTTDDALQELVELIYQQATSIPNFSYMGARLCNYLSHHLTISPQSGNFRQLLLQRCRTEYEVKDQAAKGDEVTRSpecifications
| PAIP1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_182789 | |
| PAIP1 | |
| PAIP1 (NP_001002,76 a.a. ∽ 185 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10605 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 7E7 | |
| Mouse monoclonal antibody raised against a partial recombinant PAIP1. | |
| PAIP1 | |
| MGC12360 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.