missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic exocrine cell marker, Mouse, Clone: HIC0-1C10, Abnova™
Mouse monoclonal antibody raised against pancreatic exocrine cell marker.
Brand: Abnova MAB7164.100uL
This item is not returnable.
View return policy
Specifications
| Pancreatic exocrine cell marker | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (1:50-1:100) Immunohistochemistry (Frozen sections) (1:100) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| HIC0-1C10 | |
| Mouse monoclonal antibody raised against pancreatic exocrine cell marker. | |
| In tissue culture supernatant (0.09% sodium azide) | |
| Human pancreatic enriched exocrine cells. | |
| RUO | |
| Specific for exocrine cell types. | |
| Store at 4°C. Do not freeze. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction