Learn More
PAXIP1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PAXIP1.
Brand: Abnova H00022976-A01.50uL
Description
This gene is a member of the paired box (PAX) gene family and encodes a nuclear protein with six BRCT (breast cancer carboxy-terminal) domains. This protein plays a critical role in maintaining genome stability, condensation of chromatin and progression through mitosis. [provided by RefSeq
Sequence: HLIASKVTRTVKFLTAISVVKHIVTPEWLEECFRCQKFIDEQNYILRDAEAEVLFSFSLEESLKRAHVSPLFKAKYFYITPGICPSLSTMKAIVECAGGKVLSKQPSFSpecifications
| PAXIP1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PAXIP1. | |
| PAXIP1 | |
| CAGF28/CAGF29/FLJ41049/PACIP1/PAXIP1L/PTIP/TNRC2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_007349 | |
| PAXIP1 | |
| PAXIP1 (NP_620176, 868 a.a. ∽ 975 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 22976 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only