Learn More
PBK, Mouse, Clone: 4E3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PBK.
Brand: Abnova H00055872-M05A.200uL
Description
This genes encodes a serine/threonine kinase related to the dual specific mitogen-activated protein kinase kinase (MAPKK) family. Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis. [provided by RefSeq
Sequence: MEGISNFKTPSKLSEKKKSVLCSTPTINIPASPFMQKLGFGTGVNVYLMKRSPRGLSHSPWAVKKINPICNDHYRSVYQKRLMDEAKILKSLHHPNIVGYRAFTEANDGSSpecifications
| PBK | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC015191 | |
| PBK | |
| PBK (AAH15191,1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 55872 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA | |
| 4E3 | |
| Mouse monoclonal antibody raised against a partial recombinant PBK. | |
| PBK | |
| FLJ14385/Nori-3/SPK/TOPK | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.