Learn More
PCDH8, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PCDH8.
Brand: Abnova H00005100-A01.50uL
Description
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The gene encodes an integral membrane protein that is thought to function in cell adhesion in a CNS-specific manner. Unlike classical cadherins, which are generally encoded by 15-17 exons, this gene includes only 3 exons. Notable is the large first exon encoding the extracellular region, including 6 cadherin domains and a transmembrane region. Alternative splicing yields isoforms with unique cytoplasmic tails. [provided by RefSeq
Sequence: VRYSTFEEDAPGTVIGTLAEDLHMKVSGDTSFRLMKQFNSSLLRVREGDGQLTVGDAGLDRERLCGQAPQCVLAFDVVSFSQEQFRLVHSpecifications
| PCDH8 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PCDH8. | |
| PCDH8 | |
| ARCADLIN/PAPC | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002590 | |
| PCDH8 | |
| PCDH8 (NP_002581, 32 a.a. ∽ 120 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 5100 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only