Learn More
PCSK9, Mouse, Clone: KS-2C8, Abnova™
Mouse monoclonal antibody raised against full-length recombinant PCSK9.
Brand: Abnova MAB8228.100ug
Description
This gene encodes a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. The encoded protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. This protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons. Mutations in this gene have been associated with a third form of autosomal dominant familial hypercholesterolemia (HCHOLA3). [provided by RefSeq
Specifications
| PCSK9 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) Immunoprecipitation (1-2 ug/sample) The optimal working dilution should be determined by the end user. | |
| PCSK9 | |
| PCSK9 | |
| Recombinant His fusion protein corresponding to full length human PCSK9. | |
| RUO | |
| 255738 | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| KS-2C8 | |
| Mouse monoclonal antibody raised against full length recombinant PCSK9. | |
| In 10mM phosphate buffer, 150mM NaCl, pH 7.5 (50% glycerol) | |
| FH3/HCHOLA3/LDLCQ1/NARC-1/NARC1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous mature form of PCSK9 prodomain, does not react with pro-form of PCSK9. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.