Learn More
PGAP1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PGAP1.
Brand: Abnova H00080055-A01.50uL
Description
PGAP1 catalyzes the inositol deacylation of glycosylphosphatidylinositol (GPI) at an early step in GPI biosynthesis. Inositol deacylation is essential for the generation of mature GPI capable of attachment to proteins (Tanaka et al., 2004 [PubMed 14734546]).[supplied by OMIM
Sequence: VAIIGHSMGGLVARALLTLKNFKHDLINLLITQATPHVAPVMPLDRFITDFYTTVNNYWILNARHINLTTLSVAGGFRDYQVRSGLTFLPKLSHHTSALSpecifications
| PGAP1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PGAP1. | |
| PGAP1 | |
| Bst1/FLJ42774/ISPD3024 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_024989 | |
| PGAP1 | |
| PGAP1 (NP_079265, 168 a.a. ∽ 266 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 80055 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only