missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoserine, Mouse, Clone: A8G9, Abnova™
Mouse monoclonal antibody raised against Phosphoserine.
Brand: Abnova MAB2545.100ug
This item is not returnable.
View return policy
Specifications
| Phosphoserine | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| A8G9 | |
| Mouse monoclonal antibody raised against Phosphoserine. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| A chemically linked phosphoserine. | |
| RUO | |
| This antibody recognizes phosphoserine only. It does not cross react with phosphotyrosine or phosphothreonine. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. Upon initial thawing apportion into working aliquots and store at -80°C for up to six months. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction