Learn More
PLA1A, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PLA1A.
Brand: Abnova H00051365-A01.50uL
Description
Phosphatidylserine-specific phospholipase A1-alpha (PLA1A) acts specifically on phosphatidylserine (PS) and 1-acyl-2-lysophosphatidylserine (lyso-PS) to hydrolyze fatty acids at the sn-1 position of these phospholipids.[supplied by OMIM
Sequence: DQPGCPTFFYAGYSYLICDHMRAVHLYISALENSCPLMAFPCASYKAFLAGRCLDCFNPFLLSCPRIGLVEQGGVKIEPLPKEVKVYLLTTSSAPYCMHHSpecifications
| PLA1A | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PLA1A. | |
| PLA1A | |
| PS-PLA1/PSPLA1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_015900 | |
| PLA1A | |
| PLA1A (NP_056984, 241 a.a. ∽ 340 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 51365 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only