missing translation for 'onlineSavingsMsg'
Learn More

PLA2G3, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16166366
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16166366 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16166366 Supplier Abnova Supplier No. H00050487A01

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant PLA2G3.

PLA2G3 belongs to the family of secreted phospholipase A2 (sPLA2; EC 3.1.1.4) proteins. These Ca(2+)-dependent lipolytic enzymes have a conserved Ca(2+)-binding loop and a his-asp dyad in the catalytic site (Murakami et al., 2003 [PubMed 12522102]).[supplied by OMIM

Sequence: SPALRWYRTSCHLTKAVPGNPLGYLSFLAKDAQGLALIHARWDAHRRLQACSWEDEPELTAAYGALCAHETAWGSFIHTPGPELQRALATLQSQWEACRALEESPAGARK

Specifications

Antigen PLA2G3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PLA2G3.
Formulation 50% glycerol
Gene PLA2G3
Gene Accession No. NM_015715
Gene Alias GIII-SPLA2/SPLA2III
Gene Symbols PLA2G3
Host Species Mouse
Immunogen PLA2G3 (NP_056530, 21 a.a. ∽ 130 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50487
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.