Learn More
PNMT, Mouse, Clone: AT1C11, Abnova™
Mouse monoclonal antibody raised against full-length recombinant PNMT.
Brand: Abnova MAB11111.100uL
Description
The product of this gene catalyzes the last step of the catecholamine biosynthesis pathway, which methylates norepinephrine to form epinephrine (adrenaline). The enzyme also has beta-carboline 2N-methyltransferase activity. This gene is thought to play a key step in regulating epinephrine production. [provided by RefSeq
Specifications
| PNMT | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against full length recombinant PNMT. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| MGC34570/PENT/PNMTase | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 5409 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
| ELISA, Western Blot | |
| AT1C11 | |
| Unconjugated | |
| Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
| PNMT | |
| PNMT | |
| Recombinant protein corresponding to amino acids 1-282 of human PNMT. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.