Learn More
POU3F3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant POU3F3.
Brand: Abnova H00005455-A01.50uL
Description
POU3F3 is a member of the class III POU family of transcription factors (see POU3F1; MIM 602479) that are expressed in the central nervous system. The POU domain in these proteins is required for high affinity binding to octamer DNA sequences Sumiyama et al. (1996) [PubMed 8703082].[supplied by OMIM
Sequence: AAQGRKRKKRTSIEVSVKGALESHFLKCPKPSAQEITNLADSLQLEKEVVRVWFCNRRQKEKRMTPPGIQQQTPDDVYSQVGTVSADTPPPHHGLQTSVQSpecifications
| POU3F3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant POU3F3. | |
| POU3F3 | |
| BRN1/OTF8 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006236 | |
| POU3F3 | |
| POU3F3 (NP_006227, 401 a.a. ∽ 500 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 5455 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only