Learn More
PPA2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PPA2.
Brand: Abnova H00027068-A01.50uL
Description
The protein encoded by this gene is localized to the mitochondrion, is highly similar to members of the inorganic pyrophosphatase (PPase) family, and contains the signature sequence essential for the catalytic activity of PPase. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: MSALLRLLRTGAPAAACLRLGTSAGTGSRRAMALYHTEERGQPCSQNYRLFFKNVTGHYISPFHDIPLKVNSKEENGIPMKKARNDEYENSpecifications
| PPA2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PPA2. | |
| PPA2 | |
| FLJ20459/HSPC124/MGC49850/SID6-306 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_176869 | |
| PPA2 | |
| PPA2 (NP_789845, 1 a.a. ∽ 90 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 27068 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only