missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPFIBP2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PPFIBP2.
Brand: Abnova H00008495-A01.50uL
This item is not returnable.
View return policy
Description
PPFIBP2, or liprin-beta-2, belongs to the liprin-beta family. See liprin-beta-1 (MIM 603141) for background on liprins.[supplied by OMIM
Sequence: MASDASHALEAALEQMDGIIAGTKTGADLSDGTCEPGLASPASYMNPFPVLHLIEDLRLALEMLELPQERAALLSQIPGPTAAYIKEWFEESLSQVNHHSSpecifications
| PPFIBP2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PPFIBP2. | |
| PPFIBP2 | |
| Cclp1/DKFZp781K06126/MGC42541 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003621 | |
| PPFIBP2 | |
| PPFIBP2 (NP_003612, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 8495 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction