Learn More
PPP2R2C, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PPP2R2C.
Brand: Abnova H00005522-A01.50uL
Description
The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes a gamma isoform of the regulatory subunit B55 subfamily. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: MGEDTDTRKINHSFLRDHSYVTEADIISTVEFNHTGELLATGDKGGRVVIFQREPESKNAPHSQGEYDVYSTFQSHEPEFDYLKSLEIEEKINKIKWLPQQNAAHSLLSTSpecifications
| PPP2R2C | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PPP2R2C. | |
| PPP2R2C | |
| B55-GAMMA/IMYPNO/IMYPNO1/MGC33570/PR52/PR55G | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA | |
| Unconjugated | |
| 50% glycerol | |
| NM_020416 | |
| PPP2R2C | |
| PPP2R2C (NP_065149, 1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 5522 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only