Learn More
PRKDC, Mouse, Clone: 3H6, Abnova™
Mouse monoclonal antibody raised against recombinant PRKDC.
Brand: Abnova MAB10303.100uL
Description
The PRKDC gene encodes the catalytic subunit of a nuclear DNA-dependent serine/threonine protein kinase (DNA-PK). The second component is the autoimmune antigen Ku (MIM 152690), which is encoded by the G22P1 gene on chromosome 22q. On its own, the catalytic subunit of DNA-PK is inactive and relies on the G22P1 component to direct it to the DNA and trigger its kinase activity; PRKDC must be bound to DNA to express its catalytic properties.[supplied by OMIM
Specifications
| PRKDC | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| PRKDC | |
| PRKDC | |
| Recombinant protein corresponding to human PRKDC. | |
| RUO | |
| 5591 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 3H6 | |
| Mouse monoclonal antibody raised against recombinant PRKDC. | |
| In ascites (0.03% sodium azide) | |
| DNA-PKcs/DNAPK/DNPK1/HYRC/HYRC1/XRCC7/p350 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.