missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prph, Mouse, Clone: 7C5, Abnova™
Mouse monoclonal antibody raised against recombinant Prph.
Brand: Abnova MAB3348.200uL
This item is not returnable.
View return policy
Specifications
| Prph | |
| Monoclonal | |
| Unconjugated | |
| Immunocytochemistry (1:500-1:5000) Immunofluorescence (1:500-1:5000) Western Blot (1:500-1:2500) The optimal working dilution should be determined by the end user. | |
| Prph | |
| Prph | |
| Recombinant protein corresponding to rat Prph. | |
| RUO | |
| 24688 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 7C5 | |
| Mouse monoclonal antibody raised against recombinant Prph. | |
| In PBS (50% glycerol, 0.01% thimerosal) | |
| Perf/Prph1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Chicken, Mammalia, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction