Learn More
PSA (total), Mouse, Clone: B731M, Abnova™
Mouse monoclonal antibody raised against PSA (total).
Brand: Abnova MAB6731.1mg
Description
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Its protein product is a protease present in seminal plasma. It is thought to function normally in the liquefaction of seminal coagulum, presumably by hydrolysis of the high molecular mass seminal vesicle protein. Serum level of this protein, called PSA in the clinical setting, is useful in the diagnosis and monitoring of prostatic carcinoma. Alternate splicing of this gene generates several transcript variants encoding different isoforms. [provided by RefSeq
Specifications
| PSA (total) | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| KLK3 | |
| KLK3 | |
| Purified free PSA and PSA complexed with ACT. | |
| RUO | |
| 354 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA | |
| B731M | |
| Mouse monoclonal antibody raised against PSA (total). | |
| In PBS, pH 7.4 (0.05% sodium azide) | |
| APS/KLK2A1/PSA/hK3 | |
| Mouse | |
| 1 mg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.