Learn More
PSAT1, Mouse, Clone: 1C2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PSAT1.
Brand: Abnova H00029968-M01A.200uL
Description
The protein encoded by this gene is likely a phosphoserine aminotransferase, based on similarity to proteins in mouse, rabbit, and Drosophila. Alternative splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq
Sequence: GGAAAMEKLSSIKSQTIYEIIDNSQGFYVCPVEPQNRSKMNIPFRIGNAKGDDALEKRFLDKALELNMLSLKGHRSVGGIRASLYNAVTIEDVQKLAAFMKKFLEMHQLSpecifications
| PSAT1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_058179 | |
| PSAT1 | |
| PSAT1 (NP_478059,262 a.a. ∽ 370 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 29968 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 1C2 | |
| Mouse monoclonal antibody raised against a partial recombinant PSAT1. | |
| PSAT1 | |
| EPIP/MGC1460/PSA/PSAT | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.