Learn More
PSMD1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PSMD1.
Brand: Abnova H00005707-A01.50uL
Description
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 nonidentical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a nonlysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes the largest non-ATPase subunit of the 19S regulator lid, which is responsible for substrate recognition and binding. [provided by RefSeq
Sequence: MITSAAGIISLLDEDEPQLKEFALHKLNAVVNDFWAEISESVDKIEVLYEDEGFRSRQFAALVASKVFYHLGAFEESLNYALGAGDLFNVNDNSEYVETIIAKCIDHYTKSpecifications
| PSMD1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PSMD1. | |
| PSMD1 | |
| MGC133040/MGC133041/P112/Rpn2/S1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002807 | |
| PSMD1 | |
| PSMD1 (NP_002798, 1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 5707 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only