missing translation for 'onlineSavingsMsg'
Learn More

Rat Tumors, epithelial origin, Mouse, Clone: MG1, Abnova™

Product Code. 16046736
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16046736 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16046736 Supplier Abnova Supplier No. MAB5200.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against Rat Tumors, epithelial origin.

Specifications

Antigen Rat Tumors, epithelial origin
Applications Immunohistochemistry
Classification Monoclonal
Clone MG1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Rat Tumors, epithelial origin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.05% sodium azide
Host Species Mouse
Immunogen Native purified from rat colon adenocarcinoma cells (CC531).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity MG1 is a rat strain-independent markers for tumor cells of epithelial origin, such as colon, breast, or lung cancer, etc. When injected in colon tumor-bearing rats, MG1 localizes to tumor cells (Hagenaars et al., 2001). MG1 is suitable for for immunohistochemical staining of tumor cells in frozen tissue sections as well as formalin-fixed/paraffin-embedded tissue. MG1 is also suitable for flow cytometry, Western blotting, immunoprecipitation, and ELISA. This monoclonal antibody recognizes a cell surface structure of about 80 kD expressed by rat tumor cells of epithelial origin. In immunohistochemical stainings, there is some cross reactivity in thymus, lymph nodes.
Target Species Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.